LIF444784332 Life Chemicals Inc.

3-isobutylimidazolidine-2,4-dione

  • MDL number MFCD18838903
  • Cas Number : 93099-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart