LIF444784351 Life Chemicals Inc.

2-(4-ethoxyphenyl)-2-oxoacetaldehyde hydrate

  • MDL number MFCD08705884
  • Cas Number : 1189873-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart