LIF444784360 Life Chemicals Inc.

2-methyl-1H-indol-5-ol

  • MDL number MFCD00022714
  • Cas Number : 13314-85-7
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart