LIF444784362 Life Chemicals Inc.

3-(2-chlorophenyl)morpholine hydrochloride

  • MDL number MFCD03840094
  • Cas Number : 1172507-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart