LIF444784366 Life Chemicals Inc.

1-(4-aminophenyl)-3-(4-methoxyphenyl)urea

  • MDL number MFCD09046446
  • Cas Number : 50906-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart