LIF444784370 Life Chemicals Inc.

6-(tert-butyl)-3-methylisoxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD24682941
  • Cas Number : 1356474-58-2
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart