LIF444784380 Life Chemicals Inc.

2-(5-(tert-butyl)-1,2,4-oxadiazol-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD19380108
  • Cas Number : 1244059-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart