LIF444784390 Life Chemicals Inc.

methyl 4-((3-methyl-1,2,4-oxadiazol-5-yl)methoxy)benzoate

  • MDL number MFCD20927867
  • Cas Number : 1338673-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart