LIF444784407 Life Chemicals Inc.

3-amino-N-((3-isopropyl-1,2,4-oxadiazol-5-yl)methyl)propanamide hydrochloride

  • MDL number MFCD23136105
  • Cas Number : 1435803-86-3
  • Molecular Weight : 248.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart