LIF444784409 Life Chemicals Inc.

2-amino-N-(2-(3-isopropyl-1,2,4-oxadiazol-5-yl)ethyl)acetamide hydrochloride

  • MDL number MFCD23137985
  • Cas Number : 1435908-30-7
  • Molecular Weight : 248.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart