LIF444784416 Life Chemicals Inc.

2-(3-(pyridin-3-yl)-1,2,4-oxadiazol-5-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD23136085
  • Cas Number : 1435804-37-7
  • Molecular Weight : 263.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart