LIF444784421 Life Chemicals Inc.

4-amino-N-(pyridin-4-yl)butanamide dihydrochloride

  • MDL number MFCD00453484
  • Cas Number : 1374408-07-7
  • Molecular Weight : 252.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart