LIF444784422 Life Chemicals Inc.

3-amino-5-chloro-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD12403888
  • Cas Number : 1172244-72-2
  • Molecular Weight : 256.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart