LIF444784424 Life Chemicals Inc.

3-(3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl)propan-1-amine hydrochloride

  • MDL number MFCD22201585
  • Cas Number : 1374407-72-3
  • Molecular Weight : 274.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart