LIF444784429 Life Chemicals Inc.

2-(3-(furan-2-yl)-1,2,4-oxadiazol-5-yl)ethan-1-amine hydrochloride

  • MDL number MFCD23136081
  • Cas Number : 1435804-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. RT
Availability details for
Quantity
  • Availability
  • Add to Cart