LIF444784435 Life Chemicals Inc.

3-(hydroxymethyl)-6-methoxy-1-methylquinolin-2(1H)-one

  • MDL number MFCD06738251
  • Cas Number : 114561-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart