LIF444784459 Life Chemicals Inc.

2-(2-(3-fluorophenyl)thiazolo[3,2-b][1,2,4]triazol-6-yl)ethan-1-amine

  • MDL number MFCD09883670
  • Cas Number : 1017232-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart