LIF444784464 Life Chemicals Inc.

1,2,3,4-tetrahydronaphthalene-1-carboxylic acid

  • MDL number MFCD02179127
  • Cas Number : 1914-65-4
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart