LIF444784468 Life Chemicals Inc.

2-(methylcarbamoyl)isonicotinic acid

  • MDL number MFCD18785155
  • Cas Number : 1279208-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart