LIF444784472 Life Chemicals Inc.

isoxazol-4-ylmethanamine hydrochloride

  • MDL number MFCD06738928
  • Cas Number : 173850-71-0
  • Molecular Weight : 134.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart