LIF444784473 Life Chemicals Inc.

methyl 6-(1H-pyrazol-1-yl)nicotinate

  • MDL number MFCD00140746
  • Cas Number : 321533-62-4
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart