LIF444784475 Life Chemicals Inc.

tert-butyl 3-amino-2-phenyl-2,6-dihydropyrrolo[3,4-c]pyrazole-5(4H)-carboxylate

  • MDL number MFCD24384721
  • Cas Number : 1414568-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart