LIF444784478 Life Chemicals Inc.

3-(tert-butyl)-5-methylisoxazole-4-carboxylic acid

  • MDL number MFCD07376063
  • Cas Number : 90643-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart