LIF444784480 Life Chemicals Inc.

methyl 1-methyl-5-oxo-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD22588081
  • Cas Number : 1396774-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart