LIF444784481 Life Chemicals Inc.

1,2,5-thiadiazole-3,4-diamine

  • MDL number MFCD00457948
  • Cas Number : 55904-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart