LIF444784484 Life Chemicals Inc.

(4-methyl-1,2,5-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD06739028
  • Cas Number : 1159825-48-5
  • Molecular Weight : 149.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart