LIF444784487 Life Chemicals Inc.

N-benzyl-3-nitroaniline

  • MDL number MFCD00097793
  • Cas Number : 33334-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart