LIF444784498 Life Chemicals Inc.

4-((3-(ethoxycarbonyl)-7-(trifluoromethyl)quinolin-4-yl)amino)benzoic acid

  • MDL number MFCD07808038
  • Cas Number : 793627-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart