LIF444784499 Life Chemicals Inc.

4-(2-methoxyethoxy)benzoic acid

  • MDL number MFCD07187813
  • Cas Number : 27890-92-2
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart