LIF444784500 Life Chemicals Inc.

1,1-dioxo-1lambda6-thiolane-3-carboxylic acid

  • MDL number MFCD06809748
  • Cas Number : 4785-67-5
  • Molecular Weight : 164.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart