LIF444784505 Life Chemicals Inc.

4,6,8-trimethylquinolin-2(1H)-one

  • MDL number MFCD00047596
  • Cas Number : 42414-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart