LIF444784510 Life Chemicals Inc.

6-(4-phenoxyphenyl)-4,5-dihydropyridazin-3(2H)-one

  • MDL number MFCD09221220
  • Cas Number : 39499-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart