LIF444784553 Life Chemicals Inc.

N-(3-(benzo[d]thiazol-2-yl)phenyl)-2-chloroacetamide

  • MDL number MFCD06409321
  • Cas Number : 885268-48-4
  • Molecular Weight : 302.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart