LIF444784578 Life Chemicals Inc.

3',5'-dichloro-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD01862518
  • Cas Number : 221018-04-8
  • Molecular Weight : 251.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart