LIF444784591 Life Chemicals Inc.

3-((3-chlorophenyl)sulfonamido)propanoic acid

  • MDL number MFCD05866549
  • Cas Number : 690646-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart