LIF444784628 Life Chemicals Inc.

3-(4-propoxyphenyl)propanoic acid

  • MDL number MFCD00484121
  • Cas Number : 3243-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart