LIF444784629 Life Chemicals Inc.

3-(2-chloro-6-fluorophenyl)propanoic acid

  • MDL number MFCD06660331
  • Cas Number : 88740-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart