LIF444784644 Life Chemicals Inc.

3-(3,4-dimethylphenyl)propanoic acid

  • MDL number MFCD03425686
  • Cas Number : 25173-76-6
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart