LIF444784688 Life Chemicals Inc.

2-amino-4-hydroxy-5-methoxybenzoic acid hydrochloride

  • MDL number MFCD21092080
  • Cas Number : 1351641-73-0
  • Molecular Weight : 219.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart