LIF444784689 Life Chemicals Inc.

4-(3-nitrophenoxy)benzenesulfonyl chloride

  • MDL number MFCD21092079
  • Cas Number : 1351619-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart