LIF444784724 Life Chemicals Inc.

potassium benzo[d]oxazole-2-carboxylate

  • MDL number MFCD20441325
  • Cas Number : 119130-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart