LIF448278248 Life Chemicals Inc.

6-(dimethylamino)pyridazine-3-carboxylic acid hydrochloride

  • MDL number MFCD28388530
  • Cas Number : 1820704-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart