LIF448278259 Life Chemicals Inc.

6-fluoro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD03618556
  • Cas Number : 324-03-8
  • Molecular Weight : 165.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart