LIF448278263 Life Chemicals Inc.

6-hydroxy-5-phenylnicotinic acid

  • MDL number MFCD14702021
  • Cas Number : 1214389-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. RT
Availability details for
Quantity
  • Availability
  • Add to Cart