LIF448278280 Life Chemicals Inc.

1,3-dimethyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD00159635
  • Cas Number : 25016-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart