LIF448693541 Life Chemicals Inc.

N2-cyclobutylpyridine-2,3-diamine dihydrochloride

  • MDL number MFCD29037859
  • Cas Number : 1820673-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart