LIF448693546 Life Chemicals Inc.

(S)-2-amino-4-methyl-1-(pyrrolidin-1-yl)pentan-1-one hydrochloride

  • MDL number MFCD29037829
  • Cas Number : 40847-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart