LIF448693551 Life Chemicals Inc.

1,1-difluorospiro[2.5]octan-6-one

  • MDL number MFCD28126838
  • Cas Number : 1513853-81-0
  • Molecular Weight : 160.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart