LIF448693560 Life Chemicals Inc.

2-(cyclopropylamino)ethan-1-ol

  • MDL number MFCD09815646
  • Cas Number : 35265-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart