LIF448693573 Life Chemicals Inc.

5,6,7,8-tetrahydropyrido[3,4-d]pyrimidin-2-amine dihydrochloride

  • MDL number MFCD22570201
  • Cas Number : 1820703-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart